
D-Glucose 6-phosphate sodium salt

Synonym(s):
D(+)-Glucopyranose 6-phosphate sodium salt, G-6-P Na, Robison ester
Empirical Formula (Hill Notation):
C6H12NaO9P
Products
Product Category
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing