
Deferoxamine for system suitability

Synonym(s):
Deferoxamine mesylate salt, DFOM, Deferoxamine methanesulfonate salt, Desferrioxamine mesylate salt
Empirical Formula (Hill Notation):
C25H48N6O8 · CH4O3S
CAS Number:
Molecular Weight:
656.79
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing