
Dalfampridine Related Compound C

Synonym(s):
1,3-Di(pyridin-4-yl)urea
Empirical Formula (Hill Notation):
C11H10N4O
CAS Number:
Molecular Weight:
214.22
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2