
Chlorothiazide

Synonym(s):
6-Chloro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide
Empirical Formula (Hill Notation):
C7H6ClN3O4S2
CAS Number:
Molecular Weight:
295.72
Products
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing