
Chlorobutanol hemihydrate

Synonym(s):
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, β,β,β-Trichloro-tert-butyl alcohol hemihydrate, ‘Acetone chloroform’, ‘Chlorobutanol’
Linear Formula:
Cl3CC(CH3)2OH · 0.5H2O
CAS Number:
Molecular Weight:
186.46
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing