
Cannabidiol solution

Synonym(s):
CBD, 2-[(1R,6R)-3-Methyl-6-prop-1-en-2-ylcyclohex-2-en-1-yl]-5-pentylbenzene-1,3-diol
Empirical Formula (Hill Notation):
C21H30O2
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing