
Calcium gluceptate

Synonym(s):
(2ξ)-Glucoheptonic acid calcium salt (2:1)
Empirical Formula (Hill Notation):
C14H26CaO16
CAS Number:
Molecular Weight:
490.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing