
BMP2, human

Synonym(s):
BMP-2, BMP-2A, Bone morphogenetic protein 2, Bone morphogenetic protein 2A
Products
Product Category
Brand
Biological Source
Color
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing