
Betamethasone sodium phosphate

Synonym(s):
Betamethasone 21-phosphate disodium
Empirical Formula (Hill Notation):
C22H28FNa2O8P
CAS Number:
Molecular Weight:
516.40
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2