Skip to Content
Merck
Benzene-1,4-diboronic acid ≥95.0%

Benzene-1,4-diboronic acid

Benzene-1,4-diboronic acid ≥95.0%
Synonym(s):
p-Phenylenediboronic acid, 1,4-Phenylenediboronic acid, p-Benzenediboronic acid, Benzene-1,4-diboronic acid, NSC 25410
Linear Formula:
C6H4[B(OH)2]2
CAS Number:
Molecular Weight:
165.75

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing