Skip to Content
BH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Azlon Square Shoulder Safety Wash Bottles With Driplok Vapor Venting
Products
Product Category
bottles (6)
Manufacturer
Capacity (mL)
Bottle material
Color
Vial Bottle Material
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
DWKWGW234PVENT
Azlon Square Shoulder Safety Wash Bottles With Driplok Vapor Venting,
wide-neck, low-density polyethylene bottle, orange polypropylene closure, capacity 500 mL
Expand
View Pricing
DWKWGW448VT
Azlon Square Shoulder Safety Wash Bottles With Driplok Vapor Venting,
wide-neck, low-density polyethylene bottle, orange polypropylene closure, capacity 1000 mL
Expand
View Pricing
DWKWGW446VT
Azlon Square Shoulder Safety Wash Bottles With Driplok Vapor Venting,
wide-neck, low-density polyethylene bottle, blue polypropylene closure, capacity 1000 mL
Expand
View Pricing
DWKWGW447VT
Azlon Square Shoulder Safety Wash Bottles With Driplok Vapor Venting,
wide-neck, low-density polyethylene bottle, green polypropylene closure, capacity 1000 mL
Expand
View Pricing
DWKWGW237PVENT
Azlon Square Shoulder Safety Wash Bottles With Driplok Vapor Venting,
wide-neck, low-density polyethylene bottle, red polypropylene closure, capacity 1000 mL
Expand
View Pricing
DWKWGW444VT
Azlon Square Shoulder Safety Wash Bottles With Driplok Vapor Venting,
wide-neck, low-density polyethylene bottle, yellow polypropylene closure, capacity 1000 mL
Expand
View Pricing