
Asiaticoside

Synonym(s):
Madecassol
Empirical Formula (Hill Notation):
C48H78O19
CAS Number:
Molecular Weight:
959.12
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing