
Amlodipine Related Compound A

Synonym(s):
3-Ethyl 5-methyl [2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-3,5-pyridinedicarboxylate] fumarate
Empirical Formula (Hill Notation):
C20H23ClN2O5 · C4H4O4
CAS Number:
Molecular Weight:
522.93
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2