
Acephate

Synonym(s):
O,S-Dimethyl N-acetylphosphoramidothioate
Empirical Formula (Hill Notation):
C4H10NO3PS
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2