
α,α-Diphenyl-4-piperidinemethanol

Synonym(s):
Azacyclonol
Empirical Formula (Hill Notation):
C18H21NO
CAS Number:
Molecular Weight:
267.37
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing