
3-(Trimethylsilyl)-1-propanesulfonic acid sodium salt

Synonym(s):
2,2-Dimethyl-2-silapentane-5-sulfonate sodium salt, DSS sodium salt, Sodium 3-(trimethylsilyl)-1-propanesulfonate
Linear Formula:
(CH3)3Si(CH2)3SO3Na
CAS Number:
Molecular Weight:
218.32
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2