![3-Butenoic acid 97%](/deepweb/assets/sigmaaldrich/product/structures/652/156/14f4df80-7f57-4d2c-990e-c0cd402b21b2/640/14f4df80-7f57-4d2c-990e-c0cd402b21b2.png)
3-Butenoic acid
![3-Butenoic acid 97%](/deepweb/assets/sigmaaldrich/product/structures/652/156/14f4df80-7f57-4d2c-990e-c0cd402b21b2/640/14f4df80-7f57-4d2c-990e-c0cd402b21b2.png)
Synonym(s):
Vinylacetic acid
Linear Formula:
CH2=CHCH2COOH
CAS Number:
Molecular Weight:
86.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing