Skip to Content
Merck
2-Aminomethyl-15-crown-5 95%

2-Aminomethyl-15-crown-5

2-Aminomethyl-15-crown-5 95%
Synonym(s):
1,4,7,10,13-Pentaoxacyclopentadecane-2-methanamine
Empirical Formula (Hill Notation):
C11H23NO5
CAS Number:
Molecular Weight:
249.30

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing