Skip to Content
Merck
1-Bromo-3,7-dimethyloctane 96%

1-Bromo-3,7-dimethyloctane

1-Bromo-3,7-dimethyloctane 96%
Linear Formula:
CH2BrCH2CH(CH3)CH2CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
221.18

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing