
1,1′-Carbonyl-di-(1,2,4-triazole)

Synonym(s):
CDT
Empirical Formula (Hill Notation):
C5H4N6O
CAS Number:
Molecular Weight:
164.12
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing