
1,1,3,3-Tetramethylbutyl isocyanide

Synonym(s):
tert-Octyl isocyanide, Walborsky’s reagent, 1,1,3,3-Tetramethylbutyl isocyanide
Linear Formula:
(CH3)3CCH2C(CH3)2NC
CAS Number:
Molecular Weight:
139.24
Products
Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing