Skip to Content
Merck
All Photos(9)

Key Documents

ZMS1157

Sigma-Aldrich

Anti-α-Synuclein Antibody, clone Syn211 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

NACP, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

Syn211, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone Syn211 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects -Synuclein. It targets an epitope within the C-terminal region.

Immunogen

Full-length recombinant human -Synuclein.

Application

Quality Control Testing

Evaluated by Western Blotting in Human brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected -Synuclein in Human brain tissue lysate.

Tested Applications

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected -Synuclein in Human cerebral cortex and Human Parkinson′s diseased brain tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected -Synuclein in Human cerebral cortex tissue and Human Parkinson′s diseased brain tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Alpha-synuclein (UniProt: P37840; also known as Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, NACP) is encoded by the SNCA (also known as NACP; PARK1) gene (Gene ID: 6622) in human. α-synuclein is abundantly expressed in the brain and is found in a classic amyloid fibril form within the intra-neuronal Lewy body deposits of Parkinson′s disease brains. In its monomeric form it participates in synaptic vesicle exocytosis by enhancing vesicle priming, fusion, and dilation of exocytotic fusion pores.  It acts by increasing local Ca2+ release from microdomains, which is essential for the enhancement of ATP-induced exocytosis. In its multimeric form it acts as a molecular chaperone assisting in the folding of synaptic fusion components called SNAREs at presynaptic membrane.  It is also involved in the regulation of dopamine release and transport. It is reported to reduce neuronal responsiveness to various apoptotic stimuli, leading to reduced caspase-3 activation. Post-translationally it can be phosphorylated, predominantly on serine residues. Phosphorylation of serine 129 is reported to be selective and extensive in synucleinopathy lesions. This phosphorylation is reported to promote insoluble fibril formation. Genetic alterations of SNCA gene are reported to result in aberrant polymerization into fibrils, which is associated with synucleinopathies. Mutations in SNCA gene are also known to cause Parkinson′s disease characterized by bradykinesia, resting tremor, muscular rigidity, and postural instability. The pathology of Parkinson s disease involves progressive loss of dopaminergic neurons in the substantia nigra and the presence of Lewy bodies in surviving neurons in various areas of the brain. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Huang, CC., et al. (2018).  J. Cell Sci. 131(23); jcs213017; Burre, J., et al. (2010). Science. 329(5999); 1663-1667).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

30 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service