Skip to Content
Merck
All Photos(6)

Documents

WH0004609M2

Sigma-Aldrich

Monoclonal Anti-MYC antibody produced in mouse

clone 1G7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-cMyc, Anti-v-Myc myelocytomatosis viral oncogene homolog (avian)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG3κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

MYC (MYC proto-oncogene) is a transcriptional factor and oncoprotein. c-myc is a member of MYC gene family. It is located on human chromosome 8q24. c-Myc gene codes for basic helix-loop-helix/leucine zipper (bHLH/LZ) transcription factor.
The protein encoded by this gene is a multifunctional, nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. It functions as a transcription factor that regulates transcription of specific target genes. Mutations, overexpression, rearrangement and translocation of this gene have been associated with a variety of hematopoietic tumors, leukemias and lymphomas, including Burkitt lymphoma. There is evidence to show that alternative translation initiations from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site result in the production of two isoforms with distinct N-terminal. The synthesis of non-AUG initiated protein is suppressed in Burkitt′s lymphomas, suggesting its importance in the normal function of this gene. (provided by RefSeq)

Immunogen

MYC (NP_002458, 330 a.a. ~ 439 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VRVLRQISNNRKCTSPRSSDTEENVKRRTHNVLERQRRNELKRSFFALRDQIPELENNEKAPKVVILKKATAYILSVQAEEQKLISEEDLLRKRREQLKHKLEQLRNSCA

Biochem/physiol Actions

The cellular myelocytomatosis (c-myc) oncogene plays a major role in cellular proliferation, differentiation and apoptosis. It acts as transcriptional regulator of gene expression. MYC (MYC proto-oncogene) may be essential for cancer initiation, promotion and therapy resistance. Overexpression of MYC in DLBCL (diffuse large B-cell lymphoma) results in poor outcome and invasive treatment when medicated with rituximab plus cyclophosphamide, doxorubicin, vincristine and prednisone (R-CHOP).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinicopathological significance of c-MYC in esophageal squamous cell carcinoma
Lian Y, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine (2017)
Fei Wang et al.
Science signaling, 15(719), eabd0033-eabd0033 (2022-02-02)
Social memory enables one to recognize and distinguish specific individuals. It is fundamental to social behaviors that can be mediated by the oxytocin receptor (OXTR), such as forming relationships. We investigated the molecular regulation and function of OXTR in animal
Yimeng Yin et al.
The Biochemical journal, 452(1), 67-78 (2013-02-27)
The NXF (nuclear export factor) family members are implicated in the transport of mRNA from the nucleus to the cytoplasm. Recently, some members of the NXF family have been reported to play divergent functional roles, such as post-transcriptional regulation, translational
Apoptotic signaling by c-MYC.
Hoffman B, et al.
Oncogene, 27(50), 6462-6472 (2008)
Wenjing Yan et al.
Nature communications, 14(1), 4798-4798 (2023-08-10)
UBA1 is the primary E1 ubiquitin-activating enzyme responsible for generation of activated ubiquitin required for ubiquitination, a process that regulates stability and function of numerous proteins. Decreased or insufficient ubiquitination can cause or drive aging and many diseases. Therefore, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service