Skip to Content
Merck
All Photos(1)

Documents

T5826

Sigma-Aldrich

Anti-TSG101 (N-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Tumor Susceptibility Gene 101

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 46 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

microarray: suitable
western blot: 1-2 μg/mL using cell lysates of rat fibroblast Rat-1, mouse fibroblast NIH3T3, and human A431 cells, and rat brain extract (S1 cytosolic fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSG101(7251)
mouse ... Tsg101(22088)
rat ... Tsg101(292925)

Related Categories

General description

Tumor susceptibility gene 101 protein (TSG101) is mapped to human chromosome 11p15.1. TSG101 has ubiquitin E2 variant domain.

Immunogen

synthetic peptide corresponding amino acids 2-20 located at the N-terminus of human TSG101, conjugated to KLH. This sequence is identical in rat and mouse TSG101.

Biochem/physiol Actions

Tumor susceptibility gene 101 protein (TSG101) is a component of endosomal sorting complex required for transport (ESCRT) machinery. TSG101 interacts with arrestin domain-containing protein 1 and mediates its budding through plasma membrane. It binds to ubiquitinylated proteins and conveys them for degradation by lysosomes. TSG101 is also essential for the budding human immunodeficiency virus (HIV) infected cells. TSG101 deficiency is associated with genome instability and is considered as a risk factor for tumor initiation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ubiquitin recognition by the human TSG101 protein
Sundquist WI, et al.
Molecular Cell, 13(6), 783-789 (2004)
Cell cycle-dependent subcellular localization of the TSG101 protein and mitotic and nuclear abnormalities associated with TSG101 deficiency
Xie W, et al.
Proceedings of the National Academy of Sciences of the USA, 95(4), 1595-1600 (1998)
Tsg101 and the vacuolar protein sorting pathway are essential for HIV-1 budding
Garrus JE, et al.
Cell, 107(1), 55-65 (2001)
Formation and release of arrestin domain-containing protein 1-mediated microvesicles (ARMMs) at plasma membrane by recruitment of TSG101 protein.
Nabhan JF, et al.
Proceedings of the National Academy of Sciences of the USA, 109(11), 4146-4151 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service