Skip to Content
Merck
All Photos(1)

Documents

SML0885

Sigma-Aldrich

bpV(pic)

≥95% V basis

Synonym(s):

Dipotassium Bisperoxovanadium(pic) dihydrate, Dipotassium bisperoxo (picolinato) oxovanadate (V) dihydrate, K2 [VO(O2)2C6H4NO2] · 2 H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2VOO2O2C6H4NO2 · 2H2O
CAS Number:
Molecular Weight:
367.27
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% V basis

form

powder

storage condition

protect from light

color

faintly yellow to dark yellow

solubility

H2O: 20 mg/mL, clear

storage temp.

−20°C

Application

bpV(pic) has been used in cell culture treatment to study the route of activation of multiple pathways required for HERV (human endogenous retroviruses) transcription.

Biochem/physiol Actions

bpV(pic) contains a polar side chain, and specifically inhibits PTEN(phosphatase and tensin homolog) unlike its parent compound, which targets several other phosphatases too. Thus, making bpV(pic) more a prefered inhibitor. bpV(pic) is recognized as a potent activator of multiple sclerosis (an autoimmune disorder) related human endogenous retroviruses.
bpV(pic) is a bisperoxovanadium inhibitor of protein phosphotyrosine phosphatases with selectivity for phosphatase and tensin homolog (PTEN), a tumour suppressor phosphatase involved in cell cycle regulation. IC50 values for bpV(pic) are 31 nM for PTEN compared to 12.7 μM for PTPβ and 61 μM for PTP-1β. bpV(pic) can also act as an insulin mimetic and activate insulin receptor kinase (IRK).

Other Notes

Light Sensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaodong Wu et al.
CNS neuroscience & therapeutics, 29(11), 3598-3611 (2023-06-13)
Neuronal abnormalities are closely associated with major depressive disorder (MDD). Available evidence suggests a role for microRNAs (miRNAs) in regulating the expression of genes involved in MDD. Hence, miRNAs that can be potential therapeutic targets need to be identified. A
Phosphatase and tensin homologue deleted on chromosome ten (PTEN) as a molecular target in lung epithelial wound repair.
Lai J P, et al.
British Journal of Pharmacology, 152(8), 1172-1184 (2007)
Compound signaling activates endogenous retroviruses by inducing enhancer and gene-neighborhood transcription.
Azebi S, et al.
bioRxiv, 284695 (2018)
Qian Chen et al.
Journal of inflammation research, 15, 2707-2720 (2022-05-04)
This study aims to investigate the cytoprotective and anti-inflammatory effects of an α2-adrenoreceptor (α2-AR) agonist, dexmedetomidine (Dex), on lipopolysaccharides (LPS)-induced acute lung injury and underlying mechanisms with focus on alveolar macrophage polarization modulation. C57BL/6 mice were intraperitoneally injected LPS (10
Ali A Shati et al.
Clinical and experimental pharmacology & physiology, 49(2), 275-290 (2021-09-28)
This study evaluated the protective effect of resolvin D1 (RVD1) against cadmium chloride (CdCl2 )-induced hippocampal damage and memory loss in rats and investigated if such protection is mediated by modulating the PTEN/PI3K/Akt/mTOR pathway. Adult male Wistar rats (n =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service