Skip to Content
Merck
All Photos(6)

Key Documents

HPA014404

Sigma-Aldrich

Anti-SNCG antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Breast cancer-specific gene 1 protein, Anti-Gamma-synuclein, Anti-Persyn, Anti-SR, Anti-Synoretin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GAKTKENVVQSVTSVAEKTKEQANAVSEAVVSSVNTVATKTVEEAENIAVTSGVVRKEDLRPSAPQQEGEASKEKEEVAEEAQSG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNCG(6623)

General description

The gene SNCG (synuclein γ) is a breast cancer specific gene that encodes a member of the synuclein family of proteins. SNCG mRNA is not found in normal or benign breast tissue. It is expressed at low levels in low grade in situ breast carcinoma and abundantly in advanced infiltrating carcinomas.

Immunogen

Gamma-synuclein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene SNCG (synuclein γ) serves as a marker for adverse and aggressive disease in breast cancer. The mRNA is found to be overexpressed in uterine serous carcinoma compared to uterine endometrioid adenocarcinoma. It may serve as a predicting marker for aggressive tumors and severe outcome in patients with endometrial cancer. It is also found to be expressed in several malignant ovarian tumors. It may participate in cell proliferation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72534

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Liu et al.
Breast cancer research and treatment, 62(2), 99-107 (2000-10-04)
Previously, we have shown that synuclein gamma (SNCG), a member of the brain protein synuclein family, is highly expressed in human infiltrating breast carcinomas but not expressed in normal or benign breast tissues. The SNCG mRNA was also detected in
Artur Czekierdowski et al.
Neuro endocrinology letters, 27(3), 381-386 (2006-07-04)
The synucleins are a family of small cytoplasmic proteins that are expressed predominantly in neurons. Recently, their expression has been found also in many human cancers. To understand the molecular mechanisms underlying the abnormal expression of SNCG in malignant ovarian
Paulette Mhawech-Fauceglia et al.
Gynecologic oncology, 124(1), 148-152 (2011-10-22)
Synuclein-γ (SNCG) is a marker for adverse and aggressive disease in breast cancer. In previous study, we found SNCG mRNA to be overexpressed in uterine serous carcinoma compared to uterine endometrioid adenocarcinoma. The aim of this study is to explore
Ruth Rodríguez-Barrueco et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 9(4), e2104759-e2104759 (2021-12-14)
The H19X-encoded miR-424(322)/503 cluster regulates multiple cellular functions. Here, it is reported for the first time that it is also a critical linchpin of fat mass expansion. Deletion of this miRNA cluster in mice results in obesity, while increasing the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service