Skip to Content
Merck
All Photos(1)

Documents

GE29-0915-98

Sigma-Aldrich

Superose® 6 Increase

3.2/300, Cytiva 29-0915-98, 8.6 μm avg. part. size

Synonym(s):

Size exclusion chromatography, Superose 6 column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

glass column

product line

Superose®

mol wt

5000-5000000 Da (globular proteins)

manufacturer/tradename

Cytiva 29-0915-98

parameter

<0.03 mL/min flow rate (20% ethanol at low temp)
<0.07 mL/min flow rate (20% ethanol at room temp)
<0.07 mL/min flow rate (water at low temp)
<0.15 mL/min flow rate (water at room temp)
3 Mpa (435 psi)
4-50 μL sample volume

matrix

cross-linked agarose
polymeric

matrix active group

agarose phase

avg. part. size

8.6 μm

cleaning

1-14

working range

3-12

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Superose® 6 Increase 3.2/300 is a versatile, prepacked column for size exclusion chromatography in small-scale preparative purification (4 to 50 μl sample volume), as well as for characterization and analysis of proteins and other biomolecules with molecular weights between 5 000 and 5 000 000. When high resolution is required, sample amount is limited and low buffer consumption is important, this is the column of choice.

Features and Benefits

  • Increased resolution compared with Superose 6, for higher purity and improved analysis results.
  • Reduced runtime compared with Superose 6, for rapid results.
  • Versatile use for both preparative and analytical purposes, especially for large proteins and complexes.
  • Tolerance to repeated harsh cleaning procedures at high pH, giving long column lifetime and minimal carry-over.

Suitability

Stable in all common buffers: 1 M acetic acid, 8 M urea, 6 M guanidine hydrochloride, 1% SDS, organic solvents, 0.5 M NaOH (for cleaning-in-place)

Storage and Stability

Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Superdex is a registered trademark of Cytiva
Superose is a registered trademark of Cytiva

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Understanding pressure drop in chromatography systems is crucial for optimal functionality across different parts of the system.

Protocols

Superose from Cytiva are Size Exclusion Chromatography media consisting of a composite base matrix of dextran and agarose. This page shows how to perform a separation with a superose column.

Superose from Cytiva are Size Exclusion Chromatography media consisting of a composite base matrix of dextran and agarose. This page shows how to perform a separation with a superose column.

Superose from Cytiva are Size Exclusion Chromatography media consisting of a composite base matrix of dextran and agarose. This page shows how to perform a separation with a superose column.

Superose from Cytiva are Size Exclusion Chromatography media consisting of a composite base matrix of dextran and agarose. This page shows how to perform a separation with a superose column.

See All

Related Content

Separation options for Size Exclusion Chromatography with Superose from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service