GE17-0032-01
Sephadex® G-25 Fine
Cytiva 17-0032-01, pack of 100 g
Synonym(s):
Sephadex G-25 Medium
About This Item
packaging
pack of 100 g
manufacturer/tradename
Cytiva 17-0032-01
storage condition
dry media
matrix
cross-linked dextran
particle size
>20 μm (dry)
cleaning in place
2-13
working range
2-13
suitability
suitable for bioprocess medium
Related Categories
General description
Sephadex® is a gel filtration medium prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex® differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex® G-25 is one of five different G-types ranging from G-10 for small molecules to G-75 for larger molecules. Sephadex® G-25 is available in 4 different particle sizes (Course, Medium, Fine & Superfine).
Sephadex® G-25 is available in different bulk pack sizes and convenient pre-packed formats.
As a BioProcess medium, Sephadex® G-25, meets industrial demands with security of supply and comprehensive technical and regulatory support.
Features and Benefits
- Quickly desalts, removes contaminants and transfers to a new buffer in a single step.
- Excellent recovery and minimum sample dilution
- Available in prepacked HiPrep™ Desalting columns for fast and convenient desalting
- BioProcess medium supported for industrial applications and well-established for desalting and buffer exchange in industrial applications
Storage and Stability
Analysis Note
Legal Information
replaced by
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
This page shows how to solve practical problems that may occur when running an affinity chromatography column.
Protocols
This page shows describes fractional precipitation for removing gross impurities from affinity chromatography samples with small volumes.
This page shows how to perform a separation with a Sephadex column from Cytiva which are Size Exclusion Chromatography media consisting of an epichlorohydrin cross-linked dextran.
How to perform buffer exchange and desalting with Sephadex G-25, HiTrap Desalting columns, or ÄKTAprime plus.
This page shows how to prepare samples for purification with affinity chromatography.
Related Content
Sephadex® from Cytiva and Darcy’s law in Size Exclusion Chromatography
Separation options for Size Exclusion Chromatography with Sephadex from Cytiva.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service