Skip to Content
Merck
All Photos(1)

Key Documents

ZR-75-1

87012601, human breast, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

ZR-75-1, 87012601

biological source

human breast

growth mode

Adherent

karyotype

2n = 46, modal no. 72

morphology

Epithelial

products

Not specified

receptors

Reported to express the oestrogen and progesterone receptors

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian breast carcinoma

Cell Line Description

Derived from a malignant ascitic effusion in a 63 year old female Caucasian with infiltrating ductal carcinoma. The cells are reported to express both the wildtype and variant oestrogen receptors, progesterone receptor and other steroid hormones.

Application

Hormone and tumourigenicity studies
ZR-75-1 has been used to determine the effects of cucurbitacin E (CuE) extracted from Ecballium elaterium A.Rich., Fam. Cucurbitaceae.

DNA Profile

STR-PCR Data:
Amelogenin: X
CSF1PO: 10,11
D13S317: 9
D16S539: 11
D5S818: 13
D7S820: 10,11
THO1: 7,9.3
TPOX: 8
vWA: 16,18

Culture Medium

RPMI 1640 + 2mM Glutamine + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Cells grow very slowly, do not form a confluent monolayer and attach only lightly to the substrate. After subculture, it can take up to 7 days before cells re-attach. Detachment of cells can occur in PBS and trypsinisation can produce clumps of cells. To avoid clumping do not agitate the cells by hitting or shaking the flask while waiting for the cells to detach. When subculturing, ensure the trypsin is completely removed by centrifuging the cells and resuspending them in fresh medium.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service