Skip to Content
Merck
All Photos(2)

Key Documents

268704

Sigma-Aldrich

Iron(II) sulfide

technical grade

Synonym(s):

Ferrous sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FeS
CAS Number:
Molecular Weight:
87.91
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

grade

technical grade

form

powder or chunks

concentration

60.0-72.0% Fe (titration)

density

4.84 g/mL at 25 °C (lit.)

SMILES string

S=[Fe]

InChI

1S/Fe.S

InChI key

MBMLMWLHJBBADN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Non-stoichiometric, inhomogeneous product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Finck et al.
Environmental science & technology, 46(18), 10004-10011 (2012-08-21)
The isotope (79)Se may be of great concern with regard to the safe disposal of nuclear wastes in deep geological repositories due to its long half-life and potential mobility in the geosphere. The Se mobility is controlled by the oxidation
Valerie Ronay et al.
Oral health & preventive dentistry, 9(1), 37-45 (2011-05-20)
The purpose of this review was to summarise the fundamentals about black stain, its diagnosis and possible differential diagnoses as well as its microbiology and therapy. In addition, various studies investigating the relationship between black stain and dental caries are
Wei Wang et al.
Astrobiology, 11(5), 471-476 (2011-06-29)
The iron-sulfur world (ISW) theory is an intriguing prediction regarding the origin of life on early Earth. It hypothesizes that life arose as a geochemical process from inorganic starting materials on the surface of sulfide minerals in the vicinity of
David P Summers et al.
Astrobiology, 12(2), 107-114 (2012-01-31)
Understanding the abiotic fixation of nitrogen and how such fixation can be a supply of prebiotic nitrogen is critical for understanding both the planetary evolution of, and the potential origin of life on, terrestrial planets. As nitrogen is a biochemically
Andrew D Henderson et al.
Water research, 47(3), 1267-1276 (2012-12-19)
Iron sulfide (FeS) has been extensively assessed as a reactive medium to remove both metals and halogenated organics from groundwater. However, to address its suitability as a material for permeable reactive barriers (PRBs), its propensity for solids and gas production

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service