Skip to Content
Merck
All Photos(1)

Key Documents

NIST3262

St. John′s Wort (Hypericum perforatum L.) Aerial Parts

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

powder

packaging

packet of 5 × 3.3 g

manufacturer/tradename

NIST®

General description

Other Notes

Certified for the analytes listed below.
Arsenic (As), Cadmium (Cd), Mercury (Hg)

Reference/Informational value is provided for the following analytes.
Chlorogenic Acid (C16H18O9), Rutin (C27H30O16), Hyperoside (C21H20O12), Quercitrin (C21H20O11), Hypericin (C30H16O8), Pseudohypericin (C30H16O9), Lead (Pb)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service