Skip to Content
Merck
All Photos(1)

Key Documents

IRMM471

Cementite grains in carburised pure iron

IRMM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

manufacturer/tradename

JRC

application(s)

general analytical

format

neat

storage temp.

2-8°C

General description

Certified for the C mass fraction in F3C grains.

Analysis Note

For more information please see:
IRMM471

Legal Information

IRMM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service