Skip to Content
Merck
All Photos(1)

Key Documents

95760

Millipore

Yersinia Selective Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

CIN Agar, Cefsulodin-Irgasan-Novobiocin Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.74

Agency

according to ISO 10273:2017

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12.5 g/L
crystal violet, 0.001 g/L
magnesium sulfate heptahydrate, 0.01 g/L
D-mannitol, 20 g/L
mixed peptone, 20 g/L
neutral red, 0.03 g/L
sodium chloride, 1 g/L
sodium deoxycholate, 0.5 g/L
sodium pyruvate, 2 g/L
yeast extract, 2 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages
water monitoring

microbiology

suitability

selective for Yersinia spp.

Application

For the selective cultivation of Yersinia, in particular Y. enterocolitica and Y. pseudotuberculosis from clinical samples as well as food,water etc.

Preparation Note

Dissolve 58 g in 1 litre distilled water. Sterilize by autoclaving at 121°C for 15 minutes. Cool to approx. 50°C and add 2 vial of Yersinia selective Supplement (Cat. No. 75258). Mix well and pour into petri dishes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D A Schiemann
Canadian journal of microbiology, 25(11), 1298-1304 (1979-11-01)
A new agar medium for isolation of Yersinia enterocolitica was formulated based on growth studies which defined an optimum basal, and the evaluation of selective chemical agents, dyes, and antibiotics. The final formulation, designated cefsulodin-irgasan-novobiocin(CIN) agar, provided quantitative recovery of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service