Skip to Content
Merck
All Photos(2)

Documents

90323

Supelco

Clofibric acid

analytical standard

Synonym(s):

2-(p-Chlorophenoxy)-2-methylpropionic acid, α-(p-Chlorophenoxy)isobutyric acid, Clofibric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OC(CH3)2CO2H
CAS Number:
Molecular Weight:
214.65
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

120-122 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(Oc1ccc(Cl)cc1)C(O)=O

InChI

1S/C10H11ClO3/c1-10(2,9(12)13)14-8-5-3-7(11)4-6-8/h3-6H,1-2H3,(H,12,13)

InChI key

TXCGAZHTZHNUAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agustina Manassero et al.
Environmental science and pollution research international, 22(2), 926-937 (2014-03-14)
A kinetic study of the photocatalytic degradation of the pharmaceutical clofibric acid is presented. Experiments were carried out under UV radiation employing titanium dioxide in water suspension. The main reaction intermediates were identified and quantified. Intrinsic expressions to represent the
Da-Bing Ren et al.
Journal of chromatography. A, 1355, 80-85 (2014-06-22)
Selection of an appropriate solvent system is of great importance for a successful counter-current chromatography separation. In this work, the nonrandom two-liquid (NRTL) model, a thermodynamic method, was used for predicting the partition coefficient based on a few measured partition
Zein Shaban Ibrahim et al.
Bioscience, biotechnology, and biochemistry, 78(9), 1550-1559 (2014-07-24)
Peroxisome proliferator activated receptor α (PPARα) ligands, fibrates used to control hyperlipidemia. We demonstrated CYP2B induction by clofibric acid (CFA) however, the mechanism was not clear. In this study, HepG2 cells transfected with expression plasmid of mouse constitutive androstane receptor
Julia Martín et al.
The Science of the total environment, 503-504, 97-104 (2014-06-10)
Sewage sludge disposal onto lands has been stabilized previously but still many pollutants are not efficiently removed. Special interest has been focused on pharmaceutical compounds due to their potential ecotoxicological effects. Nowadays, there is scarce information about their occurrence in
Mei-Quan Cai et al.
Environmental science and pollution research international, 22(3), 1854-1867 (2014-09-10)
A simultaneous determination method of 14 multi-class pharmaceuticals using solid-phase extraction (SPE) followed by high-performance liquid chromatography-tandem mass spectrometer (HPLC-MS/MS) was established to measure the occurrence and distribution of these pharmaceuticals in tap water and a drinking water treatment plant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service