Skip to Content
Merck
All Photos(1)

Key Documents

75258

Millipore

Yersinia Selective Supplement

suitable for microbiology

Synonym(s):

CTN Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

sterility

sterile

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Yersinia spp.

Application

An antibiotic supplement recommended for the selective isolation of Yersinia enterocolitica.

Components

(per vial, sufficient for 500 ml medium)
Cefsulodin 7.5 mg, Triclosan 2 mg, Novobiocin 1.25 mg

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service