Skip to Content
Merck
All Photos(2)

Key Documents

67107

Supelco

Ethyl palmitate

analytical standard

Synonym(s):

Ethyl hexadecanoate, Palmitic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOC2H5
CAS Number:
Molecular Weight:
284.48
Beilstein:
1782663
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.440 (lit.)

bp

192-193 °C/10 mmHg (lit.)

mp

24-26 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC

InChI

1S/C18H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h3-17H2,1-2H3

InChI key

XIRNKXNNONJFQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosa Palagano et al.
Foods (Basel, Switzerland), 9(7) (2020-07-18)
The content of fatty acid ethyl esters (FAEEs) is one of the quality parameters to define if an olive oil can be classified as extra virgin as these compounds are considered markers for virgin olive oils obtained from poor-quality olives.
Elisabetta Bertol et al.
Drug testing and analysis, 6(9), 930-935 (2014-01-17)
Fatty acid ethyl esters (FAEEs) are minor ethanol metabolites that can accumulate in hair. The performance of hair FAEEs as a biomarker that can discriminate null or moderate drinking from risky, excessive drinking was verified by evaluating the relationship between
Yong-Han Hong et al.
BioMed research international, 2014, 329712-329712 (2014-10-21)
This study aims to investigate the anti-inflammatory responses and mechanisms of Siegesbeckia orientalis ethanol extract (SOE). In cell culture experiments, RAW264.7 cells were pretreated with SOE and stimulated with lipopolysaccharide (LPS) for inflammatory mediators assay. In animal experiments, mice were
Simona Pichini et al.
Clinical chemistry and laboratory medicine, 52(8), 1179-1185 (2014-03-13)
Ethyl glucuronide (EtG) measurement in neonatal meconium has emerged as a reliable marker to objectively assess prenatal exposure to maternal ethanol complementary to fatty acid ethyl ester (FAEEs) measurement. The detection of EtG in meconium is currently a lengthy, difficult
Kuniha Konuma et al.
PloS one, 10(3), e0121528-e0121528 (2015-03-31)
Many attempts have been made to find novel therapeutic strategies for non-alcoholic steatohepatitis (NASH), while their clinical efficacy is unclear. We have recently reported a novel rodent model of NASH using melanocortin 4 receptor-deficient (MC4R-KO) mice, which exhibit the sequence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service