Skip to Content
Merck
All Photos(1)

Documents

15230

Sigma-Aldrich

[Bis(trifluoroacetoxy)iodo]benzene

purum, ≥95.0% (AT)

Synonym(s):

BTI, Iodobenzene I,I-bis(trifluoroacetate), PIFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CO2)2IC6H5
CAS Number:
Molecular Weight:
430.04
Beilstein:
764767
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥95.0% (AT)

form

solid

reaction suitability

reagent type: oxidant

mp

121-125 °C (lit.)
122-125 °C

SMILES string

FC(F)(F)C(=O)O[I](OC(=O)C(F)(F)F)c1ccccc1

InChI

1S/C10H5F6IO4/c11-9(12,13)7(18)20-17(6-4-2-1-3-5-6)21-8(19)10(14,15)16/h1-5H

InChI key

PEZNEXFPRSOYPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent for:
  • Pummerer-like reactions
  • Chemoselective deprotection of dimethoxybenzyl ethers
  • Mediating oxidative cycloisomerization
  • Tosyloxylation of anilides

Other Notes

Reagent for dehydrogenation and oxidation reactions, review; New reagent for the direct conversions of amides to amines (Hofmann-type rearrangement) and key substance for carboxyterminal peptide sequencing; Smooth dethioacetalization to the carbonyls

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.R. Almond et al.
Organic Syntheses, 66, 132-132 (1988)
G. Stork et al.
Tetrahedron Letters, 30, 287-287 (1989)
A. Varvoglis
Synthesis, 709-709 (1984)
William C Skarnes et al.
Methods (San Diego, Calif.), 164-165, 18-28 (2019-06-20)
The generation of induced pluripotent stem cell models of human disease requires efficient modification of one or both alleles depending on dominant or recessive inheritance of the disease. To faithfully recapitulate many disease variants, the introduction of a single base
Determination of asparagine and glutamine in polypeptides using bis(I,I-trifluoroacetoxy)iodobenzene.
L M Soby et al.
Analytical biochemistry, 113(1), 149-153 (1981-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service