Skip to Content
Merck
All Photos(1)

Key Documents

XX3002510

Millipore

Filter Holder Support Screen, stainless steel

Accessories for filter holders for sample preparation.

Synonym(s):

Support screen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

material

stainless steel

Quality Level

diam.

25 mm

compatibility

for use with Swinny 13

shipped in

ambient

General description

Ultraclean and dispense small volumes of solvent by squeeze-bottle action. This hand-pressure operated dispenser features a filter holder fitted with a delivery tube. Use to direct filtered solvent against surfaces.

Features & Benefits:
•Integrated hand pump eliminates the need for external pump
•Holds up to 1 L of solvent

Applications:
Solvent Filtration Prior to Contamination Analysis, Solvent Rinsing of Machined Parts and Collection Containers

XX6602500 Solvent Filtering Dispenser

Replacement Parts
XX6602501 Flask and Neoprene Stopper
XX3002502 Filter Holder Gasket, PTFE
XX3002510 Filter Holder Support Screen, stainless steel

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service