Skip to Content
Merck
All Photos(2)

Key Documents

RAWG047S6

Millipore

S-Pak® Membrane Filter

mixed cellulose esters (MCE), pore size 1.2 μm, filter diam. 47 mm, white gridded sterile membranes for microbiological analysis

Synonym(s):

Membrane filters, Semi-permeable membranes, S-Pak Filters 1.2µm 47mm white gridded sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

gridded filter
mixed cellulose esters (MCE)
white filter

Agency

ISO 7704

sterility

sterile; ethylene oxide treated

feature

easily wetted with water

manufacturer/tradename

S-Pak®

filter diam.

47 mm

pore size

1.2 μm pore size

input

sample(s) (environmental)
sample type sport drink
sample type beer
sample type soft drink
sample type cosmetics
sample type cider
sample type wine
sample type food(s)
sample type bottled water
sample type beverage(s)

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc
microbiology

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

S-Pak Filters are made from Mixed cellulose Esters (MCE) with 1.2 μm pore size, white, gridded, 47mm diameter. These sterile, ready to use, individually packed filters are easy to use and more appropriate for testing yeasts and molds in samples such as ′hard to filter′ beverages

Application

Membrane filtration method for Microbial analysis

Packaging

Individually sealed, with blue separator paper, sterile

Linkage

Replaces: RAWG 047 S0

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

S-Pak® membrane filters for microbial bioburden testing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service