Skip to Content
Merck
All Photos(1)

Key Documents

420318

Sigma-Aldrich

Kainic Acid

≥98% (HPLC), solid, amino acid receptor agonist, Calbiochem®

Synonym(s):

Kainic Acid, 2-Carboxy-4-(1-methylethenyl)-3-pyrrolidineacetic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO4
CAS Number:
Molecular Weight:
213.23
UNSPSC Code:
12352106
NACRES:
NA.77

product name

Kainic Acid, An excitatory amino acid receptor agonist selective for the kainate receptor subtype (Ki = 21 nM for ³H-kainate binding in rat striatum).

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

dilute base: 10 mg/mL
water: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C10H15NO4/c1-5(2)7-4-11-9(10(14)15)6(7)3-8(12)13/h6-7,9,11H,1,3-4H2,2H3,(H,12,13)(H,14,15)/t6-,7+,9-/m0/s1

InChI key

VLSMHEGGTFMBBZ-OOZYFLPDSA-N

General description

An excitatory amino acid receptor agonist selective for the kainate receptor subtype (Ki = 21 nM for 3H-kainate binding in rat striatum). Also activates AMPA receptors.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Kainate receptor subtype
Product does not compete with ATP.
Reversible: no
Target Ki: 21 nM for 3H-kainate binding in rat striatum

Warning

Toxicity: Harmful (C)

Other Notes

Niu, L., et al. 1996. Biochemistry35, 2030.
Watkins, J.C. and Evans, R.H. 1981. Annu. Rev. Pharmacol. Toxicol. 21, 165.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jasmin Herz et al.
Neuron, 109(22), 3609-3618 (2021-11-19)
Mechanisms governing how immune cells and their derived molecules impact homeostatic brain function are still poorly understood. Here, we elucidate neuronal mechanisms underlying T cell effects on synaptic function and episodic memory. Depletion of CD4 T cells led to memory deficits and
Zi-Yang Liu et al.
Cell & bioscience, 14(1), 66-66 (2024-05-24)
Human patients often experience an episode of serious seizure activity, such as status epilepticus (SE), prior to the onset of temporal lobe epilepsy (TLE), suggesting that SE can trigger the development of epilepsy. Yet, the underlying mechanisms are not fully
Joshua D Rieskamp et al.
iScience, 26(7), 107068-107068 (2023-08-03)
Within the adult mammalian dentate gyrus (DG) of the hippocampus, glutamate stimulates neural stem cell (NSC) self-renewing proliferation, providing a link between adult neurogenesis and local circuit activity. Here, we show that glutamate-induced self-renewal of adult DG NSCs requires glutamate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service