Skip to Content
Merck
All Photos(1)

Key Documents

1.46810

Millipore

Agarose Agar

bottle of, ready-to-use, bottle volume 250 mL , filling volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105318

Agency

EP 2.6.1
JP 4.06
USP 71
71

sterility

sterile; autoclaved

form

liquid

feature

closure type screw cap with 3 loci
ready-to-use

packaging

bottle of

technique(s)

direct inoculation: suitable

bottle capacity

250 mL

bottle volume

250 mL , filling volume

pH

7.0 ± 0.2

application(s)

cosmetics
food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

General description

Agarose Agar is a simple ready to use enrichment broth designed to allow microbial multiplication. The agar with agarose forms a gel like appearance for easy transportation which liquefies upon heating.

Application

Agarose Agar is used to multiply microbes present in test samples. Upon incubation grown microbes can be isolated and identified.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They are formulated and tested to meet the pharmacopeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, thus improving accuracy, and reducing the risk of incorrect interpretation and false results

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service