Skip to Content
Merck
All Photos(1)

Key Documents

1.00188

Millipore

Chromocult® RAMBACH Agar

ISO 6579, Specialized media for the isolation and differentiation of Salmonella spp. from food and animal feed, water and other materials.

Synonym(s):

Chromogenic Salmonella Agar, Propylene Glycol Deoxycholate Neutral Red Agar, Salmonella Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

ISO 6579
suitable for ISO

Quality Level

form

powder

technique(s)

microbiological culture: suitable

pH

7.3±0.2 (25 °C)

application(s)

food and beverages

storage temp.

15-25°C

suitability

selective for enterobacteriaceae: Salmonella spp.

General description

Chromocult® RAMBACH Agar is specifically designed for the differentiation of Salmonella species. The agar is chromogenic, providing two chromogenic substrates – propylene glycol and X-Gal ((5-bromo-4-chloro-3-indoyl- β-D-galactopyranoside).
Certain Salmonella species act on the propylene glycol producing acid which causes the pH indicator in the media to change color, and pinkish-red or crimson colonies appear. Additionally, other members of the family Enterobacteriaceae which are positive for the enzyme β-D-galactosidase cleave the substrate X-Gal and this results in the production of blue-green colonies.
Salmonella species that are positive for the β-D-galactosidase enzyme and can produce acid from propylene glycol appear as purple-violet colonies, while species that are capable of neither appear as yellow or colorless colonies.

Application

The Chromocult® RAMBACH Agar can be used for: -
  • The detection of Salmonella in foodstuff, water, animal feed
  • The differential diagnosis of Salmonella species in various samples

Analysis Note

  • The dehydrated medium appears as dark-green color.
  • The prepared medium is opaque and light pink in appearance.
  • The pH value at 25 degrees is in the range of 7.1-7.5.

Legal Information

CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
Rambach is a trademark of Balel S.A.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service