Skip to Content
Merck
All Photos(1)

Documents

LM1904

Avanti

17:0-20:4 PI(4,5)P2

Avanti Research - A Croda Brand LM1904, powder

Synonym(s):

1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phospho-(1′-myo-inositol-4′,5′-bisphosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H92N3O19P3
CAS Number:
Molecular Weight:
1084.15
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand LM1904

packaging

1 ea of 1 × (with stopper and crimp cap (LM1904-1EA))

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](OP(O)([O-])=O)[C@H](OP([O-])(O)=O)[C@@H](O)[C@H]1O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+].[NH4+]

Application

17:0-20:4 PI(4,5)P2 or 1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phospho-(1′-myo-inositol-4′,5′-bisphosphate) has been used:
  • as an internal standard in liquid chromatography tandem mass spectroscopy (LC-MS/MS) experiments
  • as a lipid standard in ultra-performance liquid chromatography-tandem mass spectrometry (UPLC/MS)
  • as a standard to measure phosphatidylinositol (4,5)P2 from fibroblasts and brain samples by MS

Biochem/physiol Actions

Phosphatidylinositol 4,5-bisphosphate (PI(4,5)P2) is critical for the phosphoinositide signaling. It acts as a primary messenger and can also as a precursor for many secondary messengers involved. PI(4,5)P2 serves as a substrate for phosphoinositide 3-kinase (PI3K) to form phosphatidylinositol 3,4,5-trisphosphate (PI3,4,5P3).

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (LM1904-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prasenjit Manna et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 35(4), 1253-1275 (2015-02-28)
Phosphatidylinositol-3,4,5-triphosphate (PtdIns(3,4,5)P₃) is one of the most important phosphoinositides and is capable of activating a wide range of proteins through its interaction with their specific binding domains. Localization and activation of these effector proteins regulate a number of cellular functions
Avishek Ghosh et al.
Bioscience reports, 39(10) (2019-10-28)
Phosphatidylinositol-5-phosphate (PI5P) is a low abundance lipid proposed to have functions in cell migration, DNA damage responses, receptor trafficking and insulin signalling in metazoans. However, studies of PI5P function are limited by the lack of scalable techniques to quantify its
Anthony M Battram et al.
The Journal of biological chemistry, 292(5), 1691-1704 (2016-12-03)
The class I PI3K family of lipid kinases plays an important role in integrin αIIbβ3 function, thereby supporting thrombus growth and consolidation. Here, we identify Ras/Rap1GAP Rasa3 (GAP1IP4BP) as a major phosphatidylinositol 3,4,5-trisphosphate-binding protein in human platelets and a key
Maria Saliakoura et al.
Oncogene, 39(14), 2948-2960 (2020-02-09)
Enhanced prostaglandin production promotes the development and progression of cancer. Prostaglandins are generated from arachidonic acid (AA) by the action of cyclooxygenase (COX) isoenzymes. However, how cancer cells are able to maintain an elevated supply of AA for prostaglandin production
Oscar Vivas et al.
Cell reports, 27(9), 2636-2648 (2019-05-30)
There is increasing evidence that the lysosome is involved in the pathogenesis of a variety of neurodegenerative disorders. Thus, mechanisms that link lysosome dysfunction to the disruption of neuronal homeostasis offer opportunities to understand the molecular underpinnings of neurodegeneration and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service