Skip to Content
Merck
All Photos(1)

Key Documents

860587P

Avanti

18:1 SM (d18:1/18:1(9Z))

Avanti Research - A Croda Brand 860587P, powder

Synonym(s):

N-oleoyl-D-erythro-sphingosylphosphorylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H81N2O6P
CAS Number:
Molecular Weight:
729.07
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860587P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860587P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCC/C=C\CCCCCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C41H81N2O6P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-41(45)42-39(38-49-50(46,47)48-37-36-43(3,4)5)40(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,32,34,39-40,44H,6-19,22-31,33,35-38H2,1-5H3,(H-,42,45,46,47)/b21-20-,34-32+/t39-,40+/m0/s1

InChI key

NBEADXWAAWCCDG-QDDWGVBQSA-N

General description

18:1 SM (d18:1/18:1(9Z)) is a version of sphingomyelin (SM) containing a C16 ceramide base and an amide-linked oleoyl (18:1) acyl chain.

Application

18:1 SM (d18:1/18:1(9Z)) may be used:
  • in liposomes to measure ostreolysin A (OlyA) binding in order to infer the effects of phosphatidylcholines (PCs) on sphingomyelin (SM)/cholesterol interactions
  • as a standard for the ratiometric quantification of SM
  • to study its effects on C2-domain mutations by surface plasmon resonance (SPR)

Packaging

2 mL Amber Glass Sealed Ampule (860587P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshinori Hirano et al.
eLife, 8 (2019-05-03)
Ca2+-stimulated translocation of cytosolic phospholipase A2α (cPLA2α) to the Golgi induces arachidonic acid production, the rate-limiting step in pro-inflammatory eicosanoid synthesis. Structural insights into the cPLA2α preference for phosphatidylcholine (PC)-enriched membranes have remained elusive. Here, we report the structure of
Ayman J Aljohani et al.
Investigative ophthalmology & visual science, 55(6), 3413-3422 (2014-05-03)
We determined the profiles of sphingomyelin, sphingoid base, sphingoid base-1-phosphate, and ceramide, and their quantitative differences between control and glaucomatous trabecular meshwork (TM) derived from human donors. Control and primary open angle glaucoma (POAG) TM samples were collected from cadaver
Elisabetta Albi et al.
International journal of molecular sciences, 15(7), 11555-11565 (2014-07-01)
Proliferating thyroid cells are more sensitive to UV-C radiations than quiescent cells. The effect is mediated by nuclear phosphatidylcholine and sphingomyelin metabolism. It was demonstrated that proton beams arrest cell growth and stimulate apoptosis but until now there have been
Shreya Endapally et al.
Cell, 176(5), 1040-1053 (2019-02-05)
Sphingomyelin and cholesterol are essential lipids that are enriched in plasma membranes of animal cells, where they interact to regulate membrane properties and many intracellular signaling processes. Despite intense study, the interaction between these lipids in membranes is not well
Delphine Milhas et al.
FEBS letters, 584(9), 1887-1894 (2009-10-28)
The plasma membrane (PM) is a major resource for production of bioactive lipids and contains a large proportion of the cellular sphingomyelin (SM) content. Consequently, the regulation of SM levels at the PM by enzymes such as sphingomyelinase (SMase) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service