Skip to Content
Merck
All Photos(2)

Key Documents

850374C

Avanti

18:1 (Δ6-Cis) PC

1,2-dipetroselenoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-dipetroselinoyl-sn-glycero-3-phosphocholine; PC(18:1(6Z)/18:1(6Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H84NO8P
CAS Number:
Molecular Weight:
786.11
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850374C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850374C

concentration

10 mg/mL (850374C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

18:1 (Δ6-Cis) PC (1,2-dipetroselenoyl-sn-glycero-3-phosphocholine) is a lipid derivative having a phosphocholine backbone. It also has two18 carbon fatty acid chains, each having one cis double bond at the 6th carbon.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

18:1 (Δ6-Cis) PC (1,2-dipetroselenoyl-sn-glycero-3-phosphocholine) may be used:
  • to generate sn-1 C18:1(Δ6-cis)- lysophosphatidylcholine (LPC) and sn-1 C18:1(Δ9-cis)-LPC standards for structural characterization of lysophosphatidylcholine (LPC) species by tandem electrospray ionization-linear ion trap-mass spectrometry (ESI-LIT-MS)
  • to study the effect on α-secretase activityin SH-SY5Y membrane
  • in liposome to study the effect of its acyl chain length and bond saturation on pore formation

Biochem/physiol Actions

Phosphatidylcholine (PC) can form a hydrophobic surface in the mucus by acting as a surfactant to inhibit the penetrance of bacteria.

Packaging

5 mL Clear Glass Sealed Ampule (850374C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammatory Bowel Disease
Integrative Medicine, 501-516 (2018)
Koldo Morante et al.
The Journal of biological chemistry, 290(17), 10850-10861 (2015-03-12)
The physicochemical landscape of the bilayer modulates membrane protein function. Actinoporins are a family of potent hemolytic proteins from sea anemones acting at the membrane level. This family of cytolysins preferentially binds to target membranes containing sphingomyelin, where they form
Marcus O W Grimm et al.
International journal of molecular sciences, 14(3), 5879-5898 (2013-03-15)
Alzheimer's disease (AD) is characterized by extracellular accumulation of amyloid-β peptide (Aβ), generated by proteolytic processing of the amyloid precursor protein (APP) by β- and γ-secretase. Aβ generation is inhibited when the initial ectodomain shedding is caused by α-secretase, cleaving
Felipe Gazos-Lopes et al.
PLoS neglected tropical diseases, 8(8), e3077-e3077 (2014-08-08)
Trypanosoma cruzi is the causative agent of the life-threatening Chagas disease, in which increased platelet aggregation related to myocarditis is observed. Platelet-activating factor (PAF) is a potent intercellular lipid mediator and second messenger that exerts its activity through a PAF-specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service