Key Documents
TEM-LC300-AU-100
Sigma-Aldrich
Lacey Carbon Supported Gold Grid
size 300 mesh, hole size 100 μm , Average, box of 25
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
hole size
100 μm , Average
size
300 mesh
General description
Lacey Carbon Supported Grid: Gold Grid, 300 mesh. Average hole size is 100 micron. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.