Skip to Content
Merck
All Photos(1)

Key Documents

QBD11217

Sigma-Aldrich

Bromoacetamido-dPEG ®3-azide

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19BrN4O4
Molecular Weight:
339.19
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry

functional group

azide
bromo

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

BrCC(NCCOCCOCCOCCN=[N+]=[N-])=O

Features and Benefits

Bromoacetamido-dPEG ®₃-azide can be used to conjugate two different proteins or an antibody with a payload by reaction of the bromoacetamido with a free thiol and by using copper-catalyzed "click" chemistry of the azide to react with terminal alkynes or strained alkynes.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service