Skip to Content
Merck
All Photos(1)

Key Documents

93370

Sigma-Aldrich

1,1,1-Tris(hydroxymethyl)propane

dist., ≥98.0% (GC)

Synonym(s):

2-Ethyl-2-hydroxymethyl-1,3-propanediol, Trimethylolpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C(CH2OH)3
CAS Number:
Molecular Weight:
134.17
Beilstein:
1698309
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.8 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥98.0% (GC)

form

(Powder or Crystals or Granules or Chunks)

autoignition temp.

1301 °F

quality

dist.

bp

159-161 °C/2 mmHg (lit.)

mp

56-58 °C (lit.)
56-61 °C

solubility

H2O: 0.1 g/mL, clear

SMILES string

CCC(CO)(CO)CO

InChI

1S/C6H14O3/c1-2-6(3-7,4-8)5-9/h7-9H,2-5H2,1H3

InChI key

ZJCCRDAZUWHFQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,1,1-Tris(hydroxymethyl)propane is used as a starting material in the synthesis of a variety of polyglycerols and polylactides. It is also used as a tripodal ligand to construct manganese-based metal-organic complexes as single molecular magnets.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

356.0 °F - Cleveland open cup

Flash Point(C)

180 °C - Cleveland open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure, and magnetic properties of a [Mn22] wheel-like single-molecule magnet.
Murugesu M, et al.
Inorganic Chemistry, 43(14), 4203-4209 (2004)
Synthesis, characterization, and viscoelastic properties of high molecular weight hyperbranched polyglycerols.
Kainthan RK, et al.
Macromolecules, 39(22), 7708-7717 (2006)
Vita Kiriliauskaitė et al.
Journal of industrial microbiology & biotechnology, 38(9), 1561-1566 (2011-02-18)
The ability of the commercial lipolytic enzyme Lipoprime 50T to catalyze the biotechnologically important synthesis of the biodegradable and environmentally acceptable trimethylolpropane (2-ethyl-2-(hydroxymethyl)-1,3-propanediol) ester of oleic acid was investigated. Simple and accurate thin-layer chromatography and computer analysis methods were used
M A Wright et al.
Applied and environmental microbiology, 59(4), 1072-1076 (1993-04-01)
A bacterium that was able to utilize Emkarate 1550 (E1550), a synthetic lubricant ester, as the sole source of carbon was isolated. The isolate was tentatively identified as Micrococcus roseus. The components of the E1550 ester, octanoate, decanoate, and 1,1,1-tris(hydroxymethyl)propane
A family of manganese rods: Syntheses, structures, and magnetic properties.
Rajaraman G, et al.
Journal of the American Chemical Society, 126(47), 15445-15457 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service