Skip to Content
Merck
All Photos(2)

Key Documents

761737

Sigma-Aldrich

6-(Trifluoromethyl)pyridine-2-carboxaldehyde

97%

Synonym(s):

6-(Trifluoromethyl)-2-pyridinecarboxaldehyde, 6-(Trifluoromethyl)picolinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4F3NO
CAS Number:
Molecular Weight:
175.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder or crystals

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cccc(C=O)n1

InChI

1S/C7H4F3NO/c8-7(9,10)6-3-1-2-5(4-12)11-6/h1-4H

InChI key

OMFVRHWVFINADV-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

167.9 °F

Flash Point(C)

75.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service