Skip to Content
Merck
All Photos(1)

Key Documents

703230

Sigma-Aldrich

1,2-Propylene-d6 carbonate

98 atom % D, 98% (CP)

Synonym(s):

4-Methyl-1,3-dioxolan-2-one-d6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D6O3
Molecular Weight:
108.13
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

liquid

bp

240 °C

density

1.258 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]C([2H])([2H])C1([2H])OC(=O)OC1([2H])[2H]

InChI

1S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3/i1D3,2D2,3D

InChI key

RUOJZAUFBMNUDX-LIDOUZCJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georgia A Pilkington et al.
Physical chemistry chemical physics : PCCP, 22(48), 28191-28201 (2020-12-10)
The effect of water on the electroactive structuring of a tribologically relevant ionic liquid (IL) when dispersed in a polar solvent has been investigated at a gold electrode interface using neutron reflectivity (NR). For all solutions studied, the addition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service